Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001123037.2 | NP_001116509.1 | 4 | 604 | |
NM_001123038.2 | NP_001116510.1 | 5 | 744 | ||
NM_013506.3 | NP_038534.2 | 5 | 744 | ||
GenBank mRNA | AK076509.1 | - | 5 | 455 | |
AK088669.1 | - | 5 | 456 | ||
AK132969.1 | - | 5 | 455 | ||
AK165898.1 | - | 4 | 315 | ||
BC094422.1 | - | 5 | 440 | 6808404 | |
CF723811.1 | - | 5 | 636 | ||
X12507.1 | - | 5 | 456 |