Gene Transcripts

Gene Symbol
Stom
Entrez Gene ID
13830
Gene Name
stomatin
Gene Aliases
Epb7.2
Chromosome Location
Chr. 2: 35313990 - 35337009 on Build GRCm38
UniGene ID
Mm.295284
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013515.2 NP_038543.1 7 2091
GenBank mRNA AK077435.1 - 7 2060
AK139242.1 - 7 2059
AK143724.1 - 7 2059
AK148975.1 - 7 2060
AK149689.1 - 7 2059
AK149821.1 - 7 2059
AK149991.1 - 7 2059
AK151129.1 - 7 2058
BC003789.1 - 6 1444 3593052
U17297.1 - 7 2095

View Full Product Details