Gene Transcripts

Gene Symbol
Zkscan6
Entrez Gene ID
52712
Gene Name
zinc finger with KRAB and SCAN domains 6
Gene Aliases
1700128E15Rik, D11Ertd714e, KOX11, Zfp535, Znf18
Chromosome Location
Chr. 11: 65807175 - 65829239 on Build GRCm38
UniGene ID
Mm.340052
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK007308.1 - 4 843
AK015125.1 - 4 819
BC043075.1 - 5 874 6404784

View Full Product Details