Gene Transcripts

Gene Symbol
Trim42
Entrez Gene ID
78911
Gene Name
tripartite motif-containing 42
Gene Aliases
4930486B16Rik
Chromosome Location
Chr. 9: 97349562 - 97369958 on Build GRCm38
UniGene ID
Mm.108347
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030219.2 NP_084495.2 3 1372
GenBank mRNA AK019654.1 - 3 1373

View Full Product Details