Gene Transcripts

Gene Symbol
Snapc1
Entrez Gene ID
75627
Gene Name
small nuclear RNA activating complex, polypeptide 1
Gene Aliases
2700033G17Rik, AU015787
Chromosome Location
Chr. 12: 73964477 - 73985925 on Build GRCm38
UniGene ID
Mm.25937
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178392.4 NP_848479.1 3, 4 523
GenBank mRNA AK012317.1 - 2, 3 180
AK138261.1 - 3, 4 524
AK152833.1 - 3, 4 529
BC030456.1 - 3, 4 505 5401615

View Full Product Details