Gene Transcripts

Gene Symbol
Slc27a4
Entrez Gene ID
26569
Gene Name
solute carrier family 27 (fatty acid transporter), member 4
Gene Aliases
BB144259, FATP4
Chromosome Location
Chr. 2: 29802680 - 29817522 on Build GRCm38
UniGene ID
Mm.330113
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011989.4 NP_036119.1 13 3116
GenBank mRNA AK033519.1 - 9 2060
AK036919.1 - 13 3118
AK155313.1 - 13 3085
AK155388.1 - 13 3086

View Full Product Details