Gene Transcripts

Gene Symbol
Myo19
Entrez Gene ID
66196
Gene Name
myosin XIX
Gene Aliases
1110055A02Rik, Myohd1
Chromosome Location
Chr. 11: 84879922 - 84911131 on Build GRCm38
UniGene ID
Mm.288689
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025414.3 NP_079690.2 23 2844
GenBank mRNA AK018609.1 - 8 874
AK080117.1 - 21 2563
BC007156.1 - 16 1760 3486558
BC046294.1 - 22 2572
BC060115.1 - 20 4289

View Full Product Details