Gene Transcripts

Gene Symbol
LCP2
Entrez Gene ID
3937
Gene Name
lymphocyte cytosolic protein 2
Gene Aliases
SLP-76, SLP76
Chromosome Location
Chr. 5: 170248084 - 170297818 on Build GRCh38
UniGene ID
Hs.304475
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005565.3 NP_005556.1 21 1936
GenBank mRNA AK122637.1 - 2 1212
AK292890.1 - 21 1897
AK296098.1 - 13 1293
AK300000.1 - 19 1779
BC016618.1 - 21 1873 4476274
U20158.1 - 21 1984

View Full Product Details