Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001164147.1 | NP_001157619.1 | 13 | 1433 | |
NM_001164148.1 | NP_001157620.1 | 13 | 1433 | ||
NM_001164149.1 | NP_001157621.1 | 13 | 1430 | ||
NM_001164150.1 | NP_001157622.1 | 13 | 1433 | ||
NM_001164151.1 | NP_001157623.1 | 13 | 1430 | ||
NM_001164152.1 | NP_001157624.1 | 13 | 1433 | ||
NM_001164153.1 | NP_001157625.1 | 13 | 1430 | ||
NM_011548.4 | NP_035678.3 | 13 | 1430 | ||
GenBank mRNA | AF352579.1 | - | 13 | 1125 | |
AK017617.1 | - | 13 | 1398 | ||
AK034777.1 | - | 13 | 1413 | ||
AK038738.1 | - | 13 | 1417 | ||
AK146786.1 | - | 13 | 1425 | ||
AK147735.1 | - | 12 | 1089 | ||
AK156265.1 | - | 13 | 1413 | ||
AK161009.1 | - | 13 | 1419 | ||
AK170674.1 | - | 13 | 1428 | ||
AK202741.1 | - | 1 | 21 | ||
BC006860.1 | - | 13 | 1384 | 2631291 | |
BC018260.1 | - | 12 | 1058 | 4187386 |