Gene Transcripts

Gene Symbol
Tbc1d17
Entrez Gene ID
233204
Gene Name
TBC1 domain family, member 17
Gene Aliases
BC017607
Chromosome Location
Chr. 7: 44840776 - 44849079 on Build GRCm38
UniGene ID
Mm.27567
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042655.1 NP_001036120.1 11 1363
GenBank mRNA AK039566.1 - 11 1178
AK046883.1 - 11 1197
AK081318.1 - 11 1176
AK083153.1 - 11 1181
AK087754.1 - 1189
AK136705.1 - 11 1188
AK149917.1 - 11 1175
AK153732.1 - 11 1175
AK170821.1 - 11 1175
BC017607.1 - 11 1363 4507589
BC035449.1 - 11 1162

View Full Product Details