Gene Transcripts

Gene Symbol
Tdpoz4
Entrez Gene ID
399675
Gene Name
TD and POZ domain containing 4
Gene Aliases
Chromosome Location
Chr. 3: 93796398 - 93797510 on Build GRCm38
UniGene ID
Mm.377924
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207272.2 NP_997155.2 1 423
GenBank mRNA BC115626.1 - 1 423
BC115627.1 - 1 419

View Full Product Details