Gene Transcripts
- Gene Symbol
- Shisa7
- Entrez Gene ID
- 232813
- Gene Name
- shisa family member 7
- Gene Aliases
- Chromosome Location
- Chr. 7: 4825551 - 4844789 on Build GRCm38
- UniGene ID
- Mm.44666
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_172737.4 |
NP_766325.3
|
3, 4 |
1086 |
|
GenBank mRNA |
AK085127.1 |
- |
2, 3 |
308 |
|
View Full Product Details