Gene Transcripts

Gene Symbol
Klhl31
Entrez Gene ID
244923
Gene Name
kelch-like 31
Gene Aliases
9830147P19Rik, D930047P17Rik, Kbtbd1
Chromosome Location
Chr. 9: 77636732 - 77660125 on Build GRCm38
UniGene ID
Mm.138238
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172925.2 NP_766513.2 2 766
GenBank mRNA AK053085.1 - 2 767
BC118027.1 - 2 765

View Full Product Details