Gene Transcripts

Gene Symbol
Tmem67
Entrez Gene ID
329795
Gene Name
transmembrane protein 67
Gene Aliases
5330408M12Rik, B230117O07, b2b1163.1Clo, b2b1291.1Clo
Chromosome Location
Chr. 4: 12039355 - 12089365 on Build GRCm38
UniGene ID
Mm.475125
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177861.4 NP_808529.2 23 2428
GenBank mRNA AK045429.1 - 23 2429
AK133059.1 - 24 2513

View Full Product Details