Gene Transcripts

Gene Symbol
Ssh1
Entrez Gene ID
231637
Gene Name
slingshot homolog 1 (Drosophila)
Gene Aliases
AW551225, Gm1394, Gm1395, SSH-1, mSSH-1L
Chromosome Location
Chr. 5: 113937096 - 113993757 on Build GRCm38
UniGene ID
Mm.389682
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198109.4 NP_932777.2 7 530
GenBank mRNA AB099287.1 - 7 565
AK089308.1 - 7 534
AK155557.1 - 7 555
AK165380.1 - 6 495
AK171210.1 - 7 579
AK171556.1 - 7 545
BC046529.1 - 1 12

View Full Product Details