Gene Transcripts
- Gene Symbol
- Dbpht2
- Entrez Gene ID
- 386753
- Gene Name
- DNA binding protein with his-thr domain
- Gene Aliases
- Spot2
- Chromosome Location
- Chr. 12: 74297474 - 74300468 on Build GRCm38
- UniGene ID
- Mm.341686
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF547221.1 |
- |
1 |
878 |
|
View Full Product Details