Gene Transcripts

Gene Symbol
Tmtc1
Entrez Gene ID
387314
Gene Name
transmembrane and tetratricopeptide repeat containing 1
Gene Aliases
Arg99, BC023818
Chromosome Location
Chr. 6: 148232430 - 148444845 on Build GRCm38
UniGene ID
Mm.137858
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198967.5 NP_945318.2 6 1140
GenBank mRNA AK046396.1 - 6 1141
AK137545.1 - 6 1112
AK162239.1 - 6 796
BC145513.1 - 4 471

View Full Product Details