Gene Transcripts

Gene Symbol
Mfap3
Entrez Gene ID
216760
Gene Name
microfibrillar-associated protein 3
Gene Aliases
2610509F16Rik, 2700079M14Rik, AW536327
Chromosome Location
Chr. 11: 57518652 - 57533817 on Build GRCm38
UniGene ID
Mm.30501
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145426.2 NP_663401.2 3 1292
NM_180599.1 NP_850930.1 3 1256
GenBank mRNA AK042988.1 - 3 1240
AK049128.1 - 1 38
AK076071.1 - 3 1258
AK155122.1 - 3 1270
BC006828.1 - 3 1251 3597878
BC058567.1 - 1 2379

View Full Product Details