Gene Transcripts

Gene Symbol
Usp1
Entrez Gene ID
230484
Gene Name
ubiquitin specific peptidase 1
Gene Aliases
Chromosome Location
Chr. 4: 98923622 - 98935543 on Build GRCm38
UniGene ID
Mm.371692
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301414.1 NP_001288343.1 3, 4 802
NM_146144.4 NP_666256.2 3, 4 783
GenBank mRNA AK080882.1 - 3, 4 784
BC018179.1 - 3, 4 663 3585191
BC020007.1 - 3, 4 635 3980469

View Full Product Details