Gene Transcripts

Gene Symbol
Mfsd4
Entrez Gene ID
213006
Gene Name
major facilitator superfamily domain containing 4
Gene Aliases
A230072B04, A930031D07Rik, AI850289
Chromosome Location
Chr. 1: 132022806 - 132068062 on Build GRCm38
UniGene ID
Mm.208445
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114662.1 NP_001108134.1 10 2226
GenBank mRNA AK085484.1 - 7 1642

View Full Product Details