Gene Transcripts

Gene Symbol
Rhbdf2
Entrez Gene ID
217344
Gene Name
rhomboid 5 homolog 2 (Drosophila)
Gene Aliases
4732465I17Rik, Rhbdl6, cub
Chromosome Location
Chr. 11: 116598165 - 116627019 on Build GRCm38
UniGene ID
Mm.89695
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167680.1 NP_001161152.1 13 1796
NM_172572.3 NP_766160.2 13 2010
GenBank mRNA AK028872.1 - 13 2011
AK154639.1 - 13 1776
AK171085.1 - 13 1792
AY167043.1 - 13 1796
BC031398.1 - 7 829
BC052182.1 - 13 1744 6335022

View Full Product Details