Gene Transcripts

Gene Symbol
PCNT
Entrez Gene ID
5116
Gene Name
pericentrin
Gene Aliases
KEN, MOPD2, PCN, PCNT2, PCNTB, PCTN2, SCKL4
Chromosome Location
Chr. 21: 46324103 - 46445769 on Build GRCh38
UniGene ID
Hs.474069
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001315529.1 NP_001302458.1 2 362
NM_006031.5 NP_006022.3 2 200
XM_005261124.4 XP_005261181.1 2 219
XM_011529594.2 XP_011527896.1 2 219
XM_017028362.1 XP_016883851.1 2 219
XM_017028363.1 XP_016883852.1 2 388
GenBank mRNA AB007862.3 - 2 363
AF515282.1 - 2 93
AK024009.1 - 2 181
BC035913.1 - 2 181
BP312870.1 - 2 200
U52962.1 - 2 145

View Full Product Details