Gene Transcripts

Gene Symbol
Pyroxd1
Entrez Gene ID
232491
Gene Name
pyridine nucleotide-disulphide oxidoreductase domain 1
Gene Aliases
Chromosome Location
Chr. 6: 142345625 - 142362630 on Build GRCm38
UniGene ID
Mm.490474
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK148411.1 - 377
BC022914.1 - 4 357
BC027061.1 - 4 363 5355217

View Full Product Details