Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026195.3 | NP_080471.2 | 4 | 343 | |
GenBank mRNA | AK010449.1 | - | 4 | 312 | |
AK010611.2 | - | 4 | 326 | ||
AK012074.1 | - | 4 | 280 | ||
AK046353.1 | - | 4 | 326 | ||
AK051020.1 | - | 321 | |||
AK088701.1 | - | 3 | 244 | ||
AK139235.1 | - | 4 | 278 | ||
AK157739.1 | - | 4 | 287 | ||
AK186275.1 | - | 1 | 32 | ||
BC039925.1 | - | 4 | 349 | 3155580 | |
CJ178786.1 | - | 4 | 343 | ||
EF437353.1 | - | 3 | 208 |