Gene Transcripts
- Gene Symbol
- Vmn1r82
- Entrez Gene ID
- 171268
- Gene Name
- vomeronasal 1 receptor 82
- Gene Aliases
- V1rg12
- Chromosome Location
- Chr. 7: 12304805 - 12305719 on Build GRCm38
- UniGene ID
- Mm.377220
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_134234.1 |
NP_598995.1
|
1 |
835 |
|
View Full Product Details