Gene Transcripts

Gene Symbol
Zbtb7c
Entrez Gene ID
207259
Gene Name
zinc finger and BTB domain containing 7C
Gene Aliases
B230208J24Rik, Kr-pok, Zbtb36
Chromosome Location
Chr. 18: 75820178 - 76148564 on Build GRCm38
UniGene ID
Mm.159332
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145356.3 NP_663331.1 5 3191
GenBank mRNA AK052470.1 - 1 729
AK156372.1 - 3 2860
BC070424.1 - 5 3191 6837447

View Full Product Details