Gene Transcripts

Gene Symbol
SPATA2
Entrez Gene ID
9825
Gene Name
spermatogenesis associated 2
Gene Aliases
PD1, PPP1R145, tamo
Chromosome Location
Chr. 20: 49903391 - 49915543 on Build GRCh38
UniGene ID
Hs.48513
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135773.1 NP_001129245.1 3 3094
NM_006038.3 NP_006029.1 3 3010
XM_006723894.1 XP_006723957.1 3 2930
XM_011529116.1 XP_011527418.1 3 2967
GenBank mRNA AB018300.1 - 3 2928

View Full Product Details