Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001286452.1 | NP_001273381.1 | 5 | 730 | |
NM_026102.3 | NP_080378.2 | 5 | 698 | ||
NM_172464.3 | NP_766052.2 | 5 | 563 | ||
GenBank mRNA | AK053785.1 | - | 5 | 619 | |
AK129185.1 | - | 5 | 541 | ||
AK147480.1 | - | 5 | 544 | ||
AK165335.1 | - | 5 | 515 | ||
AY426535.1 | - | 5 | 608 | ||
BC025638.1 | - | 4 | 460 | ||
BC038403.1 | - | 5 | 504 | ||
BC047431.1 | - | 5 | 485 | ||
BC076585.1 | - | 5 | 553 | 30622306 | |
CD352113.1 | - | 5 | 655 |