Gene Transcripts

Gene Symbol
Cggbp1
Entrez Gene ID
106143
Gene Name
CGG triplet repeat binding protein 1
Gene Aliases
AA960172, AL023996, AW045841
Chromosome Location
Chr. 16: 64851891 - 64859491 on Build GRCm38
UniGene ID
Mm.10160
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178647.2 NP_848762.1 3 929
GenBank mRNA AK049631.1 - 3 1007
AK052195.1 - 3 997
AK083450.1 - 3 934
AK086053.1 - 3 1008
BC031191.1 - 1 314
BC043097.1 - 3 929 6405954
BC057053.1 - 3 983 6827183

View Full Product Details