Gene Transcripts

Gene Symbol
Arfip2
Entrez Gene ID
76932
Gene Name
ADP-ribosylation factor interacting protein 2
Gene Aliases
2310002N04Rik, POR, Por1
Chromosome Location
Chr. 7: 105634201 - 105640416 on Build GRCm38
UniGene ID
Mm.41637
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029802.4 NP_084078.3 3 354
GenBank mRNA AK009100.1 - 3 292
AK154772.1 - 3 320
BC013794.1 - 3 265 2648132
BC022942.1 - 3 278 3583149
BC028310.1 - 3 252
BC034520.1 - 3 335 3964827
BC079603.1 - 1 850

View Full Product Details