Gene Transcripts

Gene Symbol
Tmem40
Entrez Gene ID
94346
Gene Name
transmembrane protein 40
Gene Aliases
9030407H20Rik, AV091463, W64236
Chromosome Location
Chr. 6: 115729131 - 115762466 on Build GRCm38
UniGene ID
Mm.29739
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168256.1 NP_001161728.1 10 782
NM_001168257.1 NP_001161729.1 9 707
NM_001168258.1 NP_001161730.1 5 507
NM_001168259.1 NP_001161731.1 6 274
NM_144805.2 NP_659054.2 10 650
GenBank mRNA AK078864.1 - 6 275
BC019416.1 - 10 612 5151159
BX511925.1 - 5 234
CT010303.1 - 9 580
CV561368.1 - 9 649
CX564997.1 - 10 681

View Full Product Details