Gene Transcripts

Gene Symbol
Rell1
Entrez Gene ID
100532
Gene Name
RELT-like 1
Gene Aliases
AA536743
Chromosome Location
Chr. 5: 63888667 - 63968900 on Build GRCm38
UniGene ID
Mm.243632
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145923.4 NP_666035.2 7 1128
GenBank mRNA AK133845.1 - 8 1148
AK140108.1 - 7 1128
AK159944.1 - 7 1054
AK161061.1 - 7 1054
AK198442.1 - 1 25
BC023848.1 - 3 424
BC024679.1 - 4 577
BC031198.1 - 7 1097 5059693
BC066137.1 - 7 1107 30012041
BC066160.1 - 7 1107 30012671

View Full Product Details