Gene Transcripts

Gene Symbol
Sphkap
Entrez Gene ID
77629
Gene Name
SPHK1 interactor, AKAP domain containing
Gene Aliases
4930544G21Rik, A930009L15Rik, AI852220, SKIP, mKIAA1678
Chromosome Location
Chr. 1: 83255442 - 83408388 on Build GRCm38
UniGene ID
Mm.154303
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172430.3 NP_766018.3 5 538
GenBank mRNA AK034183.1 - 6 581
AK038638.1 - 6 583
BC069832.1 - 5 504 30431760

View Full Product Details