Gene Transcripts
- Gene Symbol
- Trat1
- Entrez Gene ID
- 77647
- Gene Name
- T cell receptor associated transmembrane adaptor 1
- Gene Aliases
- C030046M14Rik, Tcrim, Trim
- Chromosome Location
- Chr. 16: 48731914 - 48810520 on Build GRCm38
- UniGene ID
- Mm.167298
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AJ297969.1 |
- |
6 |
969 |
|
AK021155.1 |
- |
1 |
559 |
|
AK138735.1 |
- |
|
1017 |
|
View Full Product Details