Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023579.4 | NP_076068.1 | 8 | 956 | |
GenBank mRNA | AF294327.1 | - | 8 | 907 | |
AK017701.1 | - | 7 | 1004 | ||
AK049422.1 | - | 8 | 928 | ||
AK053681.1 | - | 8 | 921 | ||
AK079134.1 | - | 8 | 926 | ||
AK167303.1 | - | 8 | 918 | ||
AK169664.1 | - | 8 | 924 | ||
BC020012.1 | - | 2 | 228 | ||
BC052392.1 | - | 8 | 899 | 5719784 | |
BC054814.1 | - | 8 | 1129 | 6842146 |