Gene Transcripts

Gene Symbol
Hspa12b
Entrez Gene ID
72630
Gene Name
heat shock protein 12B
Gene Aliases
2700081N06Rik
Chromosome Location
Chr. 2: 131127412 - 131145985 on Build GRCm38
UniGene ID
Mm.277465
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028306.3 NP_082582.1 13 2330
GenBank mRNA AK012548.1 - 13 2303
BC011103.1 - 13 2330 3990088

View Full Product Details