Gene Transcripts
- Gene Symbol
- MOGS
- Entrez Gene ID
- 7841
- Gene Name
- mannosyl-oligosaccharide glucosidase
- Gene Aliases
- CDG2B, CWH41, DER7, GCS1
- Chromosome Location
- Chr. 2: 74461057 - 74465410 on Build GRCh38
- UniGene ID
- Hs.516119
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
X87237.1 |
- |
4 |
1109 |
|
View Full Product Details