Gene Transcripts

Gene Symbol
Stox2
Entrez Gene ID
71069
Gene Name
storkhead box 2
Gene Aliases
4933409N07Rik, AI449080, AI835114, mKIAA1392
Chromosome Location
Chr. 8: 47180048 - 47413081 on Build GRCm38
UniGene ID
Mm.23973
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114311.1 NP_001107783.1 3 3024
NM_001286376.1 NP_001273305.1 3 3024
NM_175162.4 NP_780371.3 3 1819
GenBank mRNA AK016762.1 - 1 355
AK030015.1 - 3 1633
AK129346.1 - 3 2984
AK162847.1 - 3 1853
BC099949.1 - 3 1833 30096824
BC100572.1 - 3 1819 30085660
BC100573.1 - 3 1819 30086796
BC138237.1 - 3 1590 8861256
BC138238.1 - 3 1590 8861257
BC145211.1 - 4 1692

View Full Product Details