Gene Transcripts

Gene Symbol
Mei4
Entrez Gene ID
75033
Gene Name
meiosis-specific, MEI4 homolog (S. cerevisiae)
Gene Aliases
4930486G11Rik
Chromosome Location
Chr. 9: 81863647 - 82206007 on Build GRCm38
UniGene ID
Mm.335329
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175213.3 NP_780422.1 3 717
GenBank mRNA AK043929.1 - 3 718
AK082395.1 - 3 757
AK166209.1 - 2 507
BC096613.1 - 3 759 6469435

View Full Product Details