Gene Transcripts

Gene Symbol
Myl12b
Entrez Gene ID
67938
Gene Name
myosin, light chain 12B, regulatory
Gene Aliases
1500001M02Rik, C77744, Mylc2b, RLC-B
Chromosome Location
Chr. 17: 70973963 - 70990516 on Build GRCm38
UniGene ID
Mm.261329
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023402.2 NP_075891.1 4 867
GenBank mRNA AK002885.1 - 4 869
AK146115.1 - 4 885
AK160123.1 - 4 910

View Full Product Details