Gene Transcripts

Gene Symbol
Npc2
Entrez Gene ID
67963
Gene Name
Niemann-Pick type C2
Gene Aliases
2700012J19Rik, AA408070, AU045843, HE1
Chromosome Location
Chr. 12: 84754559 - 84773112 on Build GRCm38
UniGene ID
Mm.282556
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023409.4 NP_075898.1 3 520
GenBank mRNA AB021289.1 - 3 275
AK008603.1 - 3 307
AK009127.1 - 3 337
AK012239.1 - 3 367
AK054441.1 - 1 1315
AK151067.1 - 3 307
AK151080.1 - 1 1732
AK151133.1 - 3 308
AK152236.1 - 3 307
AK153020.1 - 3 307
AK153203.1 - 3 307
AK154838.1 - 3 307
AK158624.1 - 3 488
AK159154.1 - 3 354
AK165263.1 - 3 333
AK167869.1 - 3 308
AK170327.1 - 3 308
BC003471.1 - 3 322 2649564
BC007190.1 - 3 354 3592515

View Full Product Details