Gene Transcripts

Gene Symbol
Mboat2
Entrez Gene ID
67216
Gene Name
membrane bound O-acyltransferase domain containing 2
Gene Aliases
2810049G06Rik, AU022889, AW547221, LPCAT4, Moact2, Oact2
Chromosome Location
Chr. 12: 24831487 - 24964400 on Build GRCm38
UniGene ID
Mm.167671
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026037.3 NP_080313.2 3, 4 399
GenBank mRNA AB297383.1 - 3, 4 298
AB305046.1 - 3, 4 298
AU067514.1 - 3, 4 399

View Full Product Details