Gene Transcripts

Gene Symbol
Poc5
Entrez Gene ID
67463
Gene Name
POC5 centriolar protein homolog (Chlamydomonas)
Gene Aliases
1200014M14Rik, AV144706
Chromosome Location
Chr. 13: 96388294 - 96415587 on Build GRCm38
UniGene ID
Mm.86472
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026173.3 NP_080449.1 13 2309
GenBank mRNA AK004769.2 - 13 2237
AK098149.1 - 13 2202
AK150656.1 - 13 2310

View Full Product Details