Gene Transcripts

Gene Symbol
Ube2r2
Entrez Gene ID
67615
Gene Name
ubiquitin-conjugating enzyme E2R 2
Gene Aliases
1200003M11Rik, Cdc34b, Ubc3b
Chromosome Location
Chr. 4: 41136021 - 41193370 on Build GRCm38
UniGene ID
Mm.389540
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026275.4 NP_080551.1 4 475
GenBank mRNA AJ240086.1 - 4 434
AK003550.1 - 4 495
AK007517.1 - 4 609
AK075703.1 - 4 485
AK075714.1 - 4 476
AK182726.1 - 1 70
AK183933.1 - 1 70
AK217134.1 - 1 70
BC011112.1 - 4 483 3982808

View Full Product Details