Gene Transcripts

Gene Symbol
Nsmce1
Entrez Gene ID
67711
Gene Name
non-SMC element 1 homolog (S. cerevisiae)
Gene Aliases
2510027N19Rik
Chromosome Location
Chr. 7: 125467640 - 125491576 on Build GRCm38
UniGene ID
Mm.4467
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026330.3 NP_080606.2 5, 6 616
GenBank mRNA AK009715.1 - 5, 6 505
AK160217.1 - 5, 6 516
AK166410.1 - 5, 6 509
BC031848.1 - 5, 6 595 3984224
BC049558.1 - 5, 6 609 6707952

View Full Product Details