Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022993.3 | NP_075369.2 | 7 | 2551 | |
GenBank mRNA | AB042200.1 | - | 7 | 2453 | |
AK132169.1 | - | 2535 | |||
AK144035.1 | - | 7 | 2564 | ||
AK170325.1 | - | 7 | 2525 | ||
AK196385.1 | - | 1 | 143 | ||
AK199366.1 | - | 1 | 143 | ||
AK200549.1 | - | 1 | 143 | ||
AK206255.1 | - | 1 | 143 | ||
AK210018.1 | - | 1 | 143 | ||
AK210958.1 | - | 1 | 144 | ||
AK212606.1 | - | 1 | 143 | ||
AK214260.1 | - | 1 | 143 | ||
AK214872.1 | - | 1 | 143 | ||
AK215152.1 | - | 1 | 143 | ||
BC011058.1 | - | 3 | 831 | ||
BC052378.1 | - | 7 | 2527 | 5696275 | |
BC054447.1 | - | 7 | 2519 | 6477698 |