Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025598.2 | NP_079874.1 | 5 | 736 | |
GenBank mRNA | AK002602.1 | - | 4 | 856 | |
AK003282.1 | - | 5 | 737 | ||
AK012037.1 | - | 5 | 620 | ||
AK012327.1 | - | 5 | 738 | ||
AK012545.1 | - | 3 | 711 | ||
AK154758.1 | - | 5 | 777 | ||
AK166314.1 | - | 700 | |||
BC029225.1 | - | 4 | 672 | ||
BC058516.1 | - | 5 | 774 | 6824261 | |
BC091753.1 | - | 5 | 543 | 4218108 | |
BC100439.1 | - | 5 | 628 | 6531448 |