Gene Transcripts

Gene Symbol
Cisd2
Entrez Gene ID
67006
Gene Name
CDGSH iron sulfur domain 2
Gene Aliases
1500009M05Rik, 1500026J14Rik, 1500031D15Rik, AI848398, B630006A20Rik, Miner1, Noxp70, Zcd2
Chromosome Location
Chr. 3: 135406412 - 135424442 on Build GRCm38
UniGene ID
Mm.269993
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025902.3 NP_080178.1 2 369
GenBank mRNA AK003486.1 - 2 340
AK004257.1 - 2 340
AK005184.1 - 2 328
AK005301.1 - 2 344
AM162549.1 - 2 861
BC058279.1 - 2 369 6528467

View Full Product Details