Gene Transcripts

Gene Symbol
Psmd8
Entrez Gene ID
57296
Gene Name
proteasome (prosome, macropain) 26S subunit, non-ATPase, 8
Gene Aliases
6720456J22Rik, AA407360, AL033291, AL033322, AL033323, C76433
Chromosome Location
Chr. 7: 29174187 - 29180673 on Build GRCm38
UniGene ID
Mm.273152
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026545.3 NP_080821.3 1 354
GenBank mRNA AK002921.1 - 1 193
AK003436.1 - 1 195
AK020132.1 - 1 213
AK027973.1 - 1 158
AK051257.1 - 1 206
AK150493.1 - 1 197
AK150982.1 - 1 161
AK153329.1 - 1 195
AK159216.1 - 1 163
AK166718.1 - 1 188
AK168886.1 - 1 162
CF732286.1 - 1 354

View Full Product Details