Gene Transcripts

Gene Symbol
Tsnax
Entrez Gene ID
53424
Gene Name
translin-associated factor X
Gene Aliases
R74833, Trax
Chromosome Location
Chr. 8: 125012997 - 125034192 on Build GRCm38
UniGene ID
Mm.458292
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016909.2 NP_058605.1 3 382
GenBank mRNA AF187040.1 - 3 382
AK014437.1 - 3 330
AK030407.1 - 3 312
AK146339.1 - 3 315
AK159223.1 - 3 384
AK164453.1 - 2 157
BC004611.1 - 3 284 3585400

View Full Product Details